Learn More
World Precision Instrument Microelectrode Holder (MEH3SF)-MEH3SF10
SDP
Supplier:  World Precision Instrument MEH3SF10
Encompass
For use with these WPI Products: SYS-705, SYS-773, 767, 721, FD223
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1112