Learn More
World Precision Instrument Microelectrode Holder (MPH3)-MPH312
SDP
Supplier:  World Precision Instrument MPH312
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8678