Learn More
ACL Inc Staticide 9" X 9", 150 Microfiber wipers per bag
Supplier:  ACL Inc 8409MF
Encompass_Preferred
80% Polyester / 20% nylon ultra-low particle and fiber release, ultrasonically sealed edges, high absorbency, durablew strength in wet applications, ISO Class 5 (Class 100).
Catalog No. NC1841771