Learn More
MSC Proto Micrometer Torque Wrench, 3/8 in, 16-80 Nm, 1 ea
Supplier:  MSC 38362224
Encompass
This torque wrench is calibrated to +/- 4% in the clockwise direction and +/- 6% in the counter-clockwise direction at 20% to 100% of full scale. Calibration is performed by manually loading the torque wrenches. It is manufactured to meet standard industrial torque applications, and is suitable for general industry, heavy equipment, and maintenance.
  • Calibrated to +/- 4% clockwise direction and +/- 6% counter-clockwise direction at 20% to 100% of full scale
  • Calibration accomplished by manual loading the torque wrenches
  • Manufactured to meet standard industrial torque applications, general industry, heavy equipment, and maintenance
Catalog No. 50-191-9945