Learn More
World Precision Instrument Micromanifold, 100 µm, ID Tip
SDP
Supplier:  World Precision Instrument 502110
Encompass
The most unique feature of the MPS-2 is its perfusion micromanifold. Using the latest microfluidic techniques, the injection molded micromanifold provides the least flow resistance and dead volume of any product on the market. The flow channel inner diameter is approximately 1.0 mm, except for the last 5 mm before the junction point. This design allows a fast flow rate without using a pressured system. Small channels and a unique design at the merging point further reduce the chance of cross contamination. Dead volume is less than 100 nL at junction.
Catalog No. 50-252-1337