Learn More
Chemglass Life Sciences Chemglass Microplate, 96-Well Deep, 0.5 mL, Sterile, Round Well Top (CLS-3506-S1)
SDP
Supplier:  Chemglass Life Sciences CLS3506S1
Encompass_Preferred
The Chemglass CLS-3506-S1 microplate is a sterile, 96-well deep-well polypropylene plate with 0.5 mL well capacity, designed for sample storage, culturing, and high-throughput automated workflows.
  • 96 wells with 0.5 mL capacity each.
  • USP grade class VI polypropylene construction.
  • Round well top with conical bottom for efficient sample recovery.
  • Alphanumeric well coding for easy identification.
  • Autoclavable and compatible with SBS/ANSI automation standards.
  • Supplied without lids and nestable for space-efficient storage.
  • Sterile and suitable for biological applications.
Catalog No. 50-273-3186