Learn More
Chemglass Life Sciences Chemglass Microplate, 96-Well Deep, 1.0 mL U-Bottom (CLS-3506-S7)
SDP
Supplier:  Chemglass Life Sciences CLS3506S7
Encompass_Preferred
The CLS-3506-S7 is a 96-well deep microplate with 1.0 mL U-bottom wells designed for automated processing, sample storage, and nucleic acid workflows. The plate is manufactured from polypropylene for broad chemical compatibility and is suitable for cryogenic storage and centrifugation.
  • 96-well format compatible with automated liquid handlers and multi-channel pipettes.
  • 1.0 mL well volume for deep-well sample storage and processing.
  • U-bottom well design to facilitate aspiration and recovery.
  • Polypropylene construction for chemical resistance and low binding.
  • Autoclavable and usable at temperatures from -80°C to 120°C.
  • Rated for centrifugation up to 3,000 g.
  • Stackable design with clear alphanumeric identification.
  • Conforms to SBS/ANSI 96-well plate standards.
  • Non-pyrogenic and DNase/RNase free for molecular biology applications.
Catalog No. 50-273-3168