Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemglass Life Sciences Microplate, 96-Well Deep, 2.2 mL Wells, V Conical Bottom, Sterile (CLS-3506-S16)
SDP

Supplier:  Chemglass Life Sciences CLS3506S16

Encompass_Preferred

Deep 96-well microplate with 2.2 mL well capacity and V conical bottoms. Sterile and nestable, it is designed for sample storage, nucleic acid extraction, and integration with automated and multichannel pipetting systems.

  • 96 wells, 2.2 mL per well.
  • V conical bottom.
  • Sterile packaging.
  • Deep-well design suitable for storage and processing.
  • Temperature range: -80°C to 120°C.
  • Autoclavable.
  • Centrifuge compatible up to 3,000 g.
  • Alphanumeric well identification.
  • SBS/ANSI footprint for automation compatibility.
  • Stackable and nestable design.
  • Non-pyrogenic and DNase/RNase free.
  • Pack quantity: 5 per case (CS5).

Catalog No. 50-273-3181


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.