Learn More
Beckman Coulter Beckman Coulter Microplate carrier (set of 2) - for TS-5.1-500 swinging-bucket rotor; holds two 96-well or one deep-well plate
Supplier:  Beckman Coulter 368451
Encompass
Beckman Coulter microplate carrier (MPN 368451) is a replacement accessory supplied as a set of two. It securely holds standard and deep-well microplates for balanced centrifugation in compatible swinging-bucket benchtop rotors.
  • Supplied as a set of two carriers.
  • Holds two stacked standard 96-well microplates (maximum combined height 27.9 mm) or one deep-well plate.
  • Fits compatible swinging-bucket benchtop rotors.
  • Enables microplate centrifugation for cell culture, immunoassays, and DNA precipitation.
  • Helps ensure balanced loading and secure plate support during spins.
Catalog No. NC3036669