Learn More
Biotang Inc Digital microplate shaker(4-plate) is a high performance microplate orbital shaker which accommodates up to four microplates, from 200 to 1500 rpm, all parameters digital setting.
Supplier:  Biotang Inc MX1004A
Encompass_Preferred
Digital microplate shaker(4-plate) is a high performance microplate orbital shaker which accommodates up to four microplates, from 200 to 1500 rpm, all parameters digital setting.
Catalog No. 50-751-4965