Learn More
MilliporeSigma MICROPRESS .45UM MONITOR 48
Supplier:  MilliporeSigma MSHAWGM48
Encompass
MicropreSure Monitor Sterilized 0.45micrometer White Gridded, Filter Brand Name: MF-Millipore, Filter Surface: Gridded, Filter Color: White, Device Material: Polycarbonate, 48/PK
Catalog No. NC9202011