Learn More
ATCC MicroQuant™ Bacillus spizizenii, low CFU (ATCC 6633-LQ-PACK™)
Supplier:  ATCC 6633LQPACK
Encompass
MicroQuant™ Bacillus spizizenii (low CFU) is a quantitated, best-in-class control developed to support USP General Chapter <61>. This ISO 17034 reference material is provided as a pack containing 5 vials of cryopreserved pellets (100 to 1,000 CFU per vial) and 5 vials of rehydration buffer. This convenient, single-use format rehydrates immediately and is easy to use and stored at 2–8°C. The strain is recommended for use in compendial testing.
Biosafety Level: BSL-1
Additional recommended strains for USP <61>:
P. paraeruginosa (NC3456065, ATCC 9027-LQ-PACK™)
S. aureus subsp. aureus (NC3456072, ATCC 6538-LQ-PACK™)
C. albicans (NC3456089, ATCC 10231-LQ-PACK™)
A. brasiliensis (NC3456088, ATCC 16404-LQ-PACK™)
Find more MicroQuant™ products
Catalog No. NC3456053