Learn More
ATCC MicroQuant™ Salmonella enterica, high CFU (ATCC 14028-HQ-PACK™)
Supplier:  ATCC 14028HQPACK
Encompass
"MicroQuant™ Salmonella enterica (high CFU) is a quantitated, best-in-class control. This ISO 17034 reference material is provided as a pack containing 5 vials of cryopreserved pellets (107 to 108 CFU per vial) and 5 vials of rehydration buffer. This convenient, single-use format rehydrates immediately and is easy to use and stored at 2–8°C. The strain is recommended for use in compendial testing and is not intended for therapeutic or diagnostic applications.
Biosafety Level: BSL-2
Find more MicroQuant™ products"
Catalog No. NC3853593