Learn More
ATCC MicroQuant™ Staphylococcus aureus subsp. aureus, low CFU (ATCC 6538-LQ-PACK™)
Supplier:  ATCC 6538LQPACK
Encompass
MicroQuant™ Staphylococcus aureus subsp. aureus (low CFU) is a quantitated, best-in-class control developed to support USP General Chapter <61>. This ISO 17034 reference material is provided as a pack containing 5 vials of cryopreserved pellets (100 to 1,000 CFU per vial) and 5 vials of rehydration buffer.
Biosafety Level: BSL-2
Additional recommended strains:
B. cepacia (NC3796837, ATCC 25416-LQ-PACK™)
B. cenocepacia (NC3796842, ATCC BAA-245-LQ-PACK™)
B. multivorans (NC3796844, ATCC BAA-247-LQ-PACK™)
P. paraeruginosa (NC3456065, ATCC 9027-LQ-PACK™)
A. brasiliensis (NC3456088, ATCC 16404-LQ-PACK™)
B. spizizenii (NC3456053, ATCC 6633-LQ-PACK™)
E. coli (ATCC 8739-LQ-PACK™)
Find more MicroQuant™ products
Catalog No. NC3456072