Learn More
ATCC MicroQuant™ Staphylococcus aureus subsp. aureus, high CFU (ATCC 6538-HQ-PACK™)
Supplier:  ATCC 6538HQPACK
Encompass
MicroQuant™ Staphylococcus aureus subsp. aureus (high CFU) is a quantitated, best-in-class control. This ISO 17034 reference material is provided as a pack containing 5 vials of cryopreserved pellets (107 to 108 CFU per vial) and 5 vials of rehydration buffer. This convenient, single-use format rehydrates immediately and is easy to use and stored at 2–8°C. The strain is recommended for use in compendial testing and is not intended for therapeutic or diagnostic applications.
Biosafety Level: BSL-2
Additional recommended strains USP <51>:
E. coli (NC3456067, ATCC 8739-HQ-PACK™)
P. paraeruginosa (NC3456054, ATCC 9027-HQ-PACK™)
C. albicans (NC3456076, ATCC 10231-HQ-PACK™)
A. brasiliensis (NC3456077, ATCC 16404-HQ-PACK™)
Find more MicroQuant™ products
Catalog No. NC3456070