Learn More
Bulbman Inc ESB Microscope Light Bulb, 6 Volt, 20 Watt
SDP
Supplier:  Bulbman Inc 103621
Encompass_Preferred
ESB Light Bulb, 6V, 20w, 0.4 in. dia., Base: G4, Overall Length: 1.2 in., Life: 100 hr.
Catalog No. NC9427264