Learn More
Bio Rad Laboratories MICROSEAL B ADHESV SEAL 100/PK
Supplier:  Bio Rad Laboratories MSB1001
Encompass
Seal; Bio-Rad; B adhesive; Strong adhesive for reliable sealing; Optically clear for real-time PCR; Robust adhesive effective from -40 deg.C to 110 deg.C; 100
Catalog No. 50-901-0045
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product