Learn More
Sigma Aldrich Fine Chemicals Biosciences MicrospinTM G 50 Columns C
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE27533001
Encompass
MicroSpin(TM) G-50 columns are designed for the rapid purification of DNA for use in a wide range of applications including desalting buffer exchange removal of dye terminators from cycle sequencing reactions and removal of labelled nucleotides from DNA labelling reactions. Good product yield and purity is obtained with sample volumes from 12-50 ul. They are suitable for any DNA greater than 20 bases in length. They will not remove or denature enzyme.
Catalog No. 50-290-4932