Learn More
Electron Microscopy Sciences 81-Well Storage Rack, Color: Black., 5/PK
SDP
Supplier:  Electron Microscopy Sciences 61600BLK
Encompass_Preferred
With an easy open, friction fit lid, these boxes are safe for low temperature storage. Designed for quick visual identification, the uniform wells have a location ID and the tube capacity indicated on the outside of the box. The boxes are debossed with a distinctive wave design and have a frosted labeling area. Capacity for 81 wells that hold 1.5 / 2.0 ml tubes. Color: Black.
Catalog No. 50-209-0618