Learn More
Alkali Scientific Microtube with Cap, 1.5mL, Opaque Black, 500/cs
SDP
Supplier:  Alkali Scientific C3000OB
Encompass
Alkali Scientific 1.5mL Microtubes are made of premium polypropylene opaque black plastic which allows for high centrifugal strength and protection from light. They feature a secure-snap cap that ensures a strong seal and eliminates leakage and evaporation. Capable of withstanding boiling temperatures up to 121° C and freezing down to -80° C, the Qwik-Snapª micro centrifuge tubes are the perfect choice for your research.
Catalog No. 50-238-9369