Learn More
ATCC Microvascular Endothelial Cell Growth Kit-BBE
Supplier:  ATCC PCS110040
Encompass
Microvascular Endothelial Cell Growth Kit-BBE contains components that when added to Vascular Cell Basal Medium (ATCC PCS-100-030) create a complete cell culture environment for microvascular endothelial cells isolated from normal human tissues.
Catalog No. 50-238-4173