Learn More
AIR CONTROL INC Microvoid FH-25 Process Hood
SDP
Supplier:  AIR CONTROL INC FH25PVC6FT
Encompass_Preferred
Microvoid Model FH-25-Type I PVC Wet Process Hoods are constructed in flame-retardant UL 1805-compliant Type I PVC Other construction materials also include polypropylene (PP), CP7D, 304 stainless steel (SS), with 316L work deck segment. Corrosion resistant polypropylene or stainless steel construction makes it well-suited for applications involving strong acids, bases, and many solvents. The FH-25 features front located valve and instrument panels for easy access and servicing. The vertically sliding sash offers full operator protection and continuous easy access to the hood interior, A complete line of standard and custom process tanks are available. 48, 60, 72, 96, or 120 inches widths available, 35-3/8 inches deep, and 83 inches high over all (including a 6-in high removable exhaust collar(s). Segmented work surface is 27-in deep and 36-in above the floor with front located spill containment lip. MADE IN USA.
Catalog No. NC3311142
Add to Cart
Request a Quote