Learn More
Quidel Corporation MICROVUE BA ELISA KIT
SDP
Supplier:  Quidel Corporation A033
Encompass
The MicroVue Ba Enzyme Immunoassay Kit measures the amount of the complement fragment Ba in human urine, plasma or serum.
Catalog No. NC2095025