Learn More
Quidel Corporation MicroVue TRAP5b enzyme Assay kit
SDP
Supplier:  Quidel Corporation 8033
Encompass
The MicroVue TRAP5b Assay is an immunocapture enzyme assay for the determination of tartrate-resistant acid phosphatase isoform 5b (TRAcP 5b) in human serum and plasma.
Catalog No. NC2207194