Learn More
ABclonal Technology MICU2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A12198100UL
Encompass_Preferred
Enables protein heterodimerization activity. Involved in calcium import into the mitochondrion and negative regulation of mitochondrial calcium ion concentration. Located in mitochondrial inner membrane and mitochondrial intermembrane space. Part of uniplex complex.
Catalog No. 50-001-30944