Learn More
Aobious Inc MID-35, 1mg Chemical Name: Myostatin inhibitor D-peptide; D-Leu-D-Arg-D-Chg-D-Lys-D-Arg-D-Trp-D-Ile-D-Arg-D-Chg-D-Lys-D-Ile-D-Trp-D-Arg-D-Ile-D-Tyr-D-Trp-NH2
Supplier:  Aobious Inc AOB16452
Encompass
Molecular Formula: C118H184N34O17, Molecular Weight: 2350.99, Storage Condition: 0°C (short term), -20°C (long term), desiccated, Solubility: DMSO, Purity: 99% (HPLC), Synonym: Myostatin inhibitor D-peptide
Catalog No. NC3595590