Learn More
Medchemexpress LLC Midaglizole | 66529-17-7 | 98.3% | 251.33 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1485295MG
Encompass_Preferred
Midaglizole is a potent α2-adrenoceptor antagonist and a hypoglycemic agent. It is used in research and has been shown to increase blood pressure and reduce blood glucose levels in vivo.
  • Potent alpha2-adrenoceptor antagonist.
  • Hypoglycemic agent.
  • Increases blood pressure in vivo.
  • Reduces blood glucose levels in vivo.
  • For research use only.
Catalog No. 50-003-61150