Learn More
Medchemexpress LLC Midaglizole hydrochloride | 79689-25-1 | MFCD28048171 | 324.25 | 25 MG
Supplier: Medchemexpress LLC 25MGHYU00165
Midaglizole hydrochloride is a potent α2-adrenoceptor antagonist and a hypoglycemic agent. It is known to increase blood pressure and reduce blood glucose levels in vivo through various mechanisms, including the stimulation of insulin release.
- Potent α2-adrenoceptor antagonist
- Hypoglycemic agent
- Stimulates insulin release
- Increases blood pressure in rats
- Reduces blood glucose levels in dogs