Learn More
Medchemexpress LLC MIDD0301 (synonyms: GL-II-93) | 2187489-08-1 | 98.1% | 414.23 | C19H13BrFN3O2 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12380125MG
Encompass_Preferred
MIDD0301 (GL-II-93) is an orally active γ-aminobutyric acid type A (GABAAR) receptor ligand investigated for bronchodilatory and anti-asthmatic effects in preclinical studies. It relaxes histamine-contracted airway smooth muscle and exhibits minimal systemic immunotoxicity in repeat-dose evaluations, making it useful as a research reagent in respiratory and neuropharmacology.
Catalog No. 50-002-36832