Learn More
LUNA NANOTECH INC Midi Silica Spin Columns, 20 Columns
Supplier:  LUNA NANOTECH INC USP00620N
Encompass_Preferred
PuroSPIN™ MIDI Spin Columns for DNA / RNA Extraction and Purification, 20 columns
Have you ever ran out of columns that come with your favorite plasmid extraction kit but still had a lot of left-over buffer reagents remaining? Instead of buying a new expensive kit you can use PuroSPIN™ columns with the left-over buffers as a cost-effective alternative. Simply use PuroSPIN™ columns while following the manufacturer-specific protocols.
Nucleic acid binding capacity: 1.2 mg of genomic DNA, 200 µg of plasmid DNA
Compatible as cost-effective replacement columns for Plasmid DNA purification kits from Qiagen, ThermoFisher, Bio-Rad, Promega and other manufacturers
Each item includes both the spin column and a sample collection tube
Extraction of genomic DNA from bacteria, yeast, cultured cells, blood, and animal tissue
Catalog No. 50-256-6905