Learn More
REPLIGEN CORPORATION Repligen MidiKros Hollow Fiber Module, 100 kD MWCO, Sterile, 20 cm
SDP
Supplier:  REPLIGEN CORPORATION D02E10005S
Encompass
The Repligen MidiKros Hollow Fiber Filter Module is a sterile, high-efficiency unit designed for crossflow membrane separation, featuring a 100 kD MWCO and mPES membrane. This module ensures precise separation of small molecules and particles, offering optimal flow rates for enhanced productivity in scientific laboratories. Its robust construction facilitates seamless integration into existing systems, making it stand out for reliable performance in critical applications.
Catalog No. NC2282257