Learn More
Medchemexpress LLC Midodrine-d6 hydrochloride | 1188265-43-1 | 99.4% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12749AS10MG
Encompass_Preferred
Midodrine-d6 hydrochloride is a selective and orally active adrenergic α1-receptor agonist. It strengthens vascular contraction and is used for research related to cardiovascular diseases, such as orthostatic hypotension. It can also be used as a tracer or an internal standard for quantitative analysis.
  • Selective and orally active adrenergic α1-receptor agonist
  • Strengthens vascular contraction
  • Used in research for cardiovascular diseases
  • Can be used as a tracer or internal standard
  • Increases p-AMPK and PPARδ expression in certain cells
  • Increases Ca2+ in certain cells
  • Increases mitochondrial oxidative phosphorylation and ATP content in C2C12 cells
  • Reduces cellular lipid content in 3T3-L1 cells
  • Exhibits anti-hypotensive effects in orthostatic hypotension rat models
  • Relieves hypertension in spontaneously hypertensive rat models
Catalog No. 50-003-39419