Learn More
TARGETMOL CHEMICALS INC MIDODRINE HYDROCHLORIDE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T157725MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Midodrine is converted to its active metabolite desglymidodrine by deglycination reaction. Desglymidodrine selectively binds to and activates alpha-1-adrenergic receptors of the arteriolar and venous vasculature. This causes smooth muscle contraction and leads to an elevation of blood pressure. Desglymidodrine diffuses poorly across the blood-brain barrier and is therefore not associated with effects on the central nervous system. Midodrine hydrochloride (Pro-Amatine) is the hydrochloride salt form of midodrine a direct-acting prodrug and sympathomimetic agent with antihypotensive properties. purity: 99%
Catalog No. 50-257-7349