Learn More
Cayman Chemical MIdodrInhydrochlorIde 100mg
SDP
Supplier:  Cayman Chemical 17349100 MG
Encompass_Preferred
A prodrug form of desglymidodrine; increases MAP and decreases heart rate in normotensive rats at 5 mg/kg
Catalog No. 50-296-7177