Learn More
Cayman Chemical MIdodrInhydrochlorIde 250mg
SDP
Supplier:  Cayman Chemical 17349250 MG
Encompass_Preferred
A prodrug form of desglymidodrine; increases MAP and decreases heart rate in normotensive rats at 5 mg/kg
Catalog No. 50-296-6492