Learn More
TARGETMOL CHEMICALS INC MIDOSTAURIN 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T321110MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PKC412(Midostaurin (PKC412); CGP41231; CGP41251) is a broad spectrum protein kinase inhibitor. Midostaurin inhibits protein kinase C alpha (PKCalpha) vascular endothelial growth factor receptor 2 (VEGFR2) c-kit platelet-derived growth factor receptor (PDGFR) and FMS-like tyrosine kinase 3 (FLT3) tyrosine kinases which may result in disruption of the cell cycle inhibition of proliferation apoptosis and inhibition of angiogenesis in susceptible tumors. purity: 99%
Catalog No. 50-257-6973