Learn More
Medchemexpress LLC Midostaurin | 120685-11-2 | 99.81% | 570.64 | 50 MG
SDP
Supplier:  Medchemexpress LLC 50MGHY10230
Encompass_Preferred
Midostaurin (PKC412; CGP 41251) is an orally active, reversible multi-targeted protein kinase inhibitor. It inhibits PKCα/β/γ, Syk, Flk-1, Akt, PKA, c-Kit, c-Fgr, c-Src, FLT3, PDFRβ, and VEGFR1/2 with IC50s ranging from 22-500 nM. Midostaurin also upregulates endothelial nitric oxide synthase (eNOS) gene expression and demonstrates powerful anticancer effects. It is intended for research use only.
  • Orally active and reversible
  • Multi-targeted protein kinase inhibitor
  • Up-regulates endothelial nitric oxide synthase (eNOS) gene expression
  • Demonstrates powerful anticancer effects
  • Used for research and analytical applications
Catalog No. 50-003-65943