Learn More
Medchemexpress LLC Mif-1 TFA (Pro-Leu-Gly-NH2 TFA) | 35240-69-8 | 99.3% | 398.38 | C15H25F3N4O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY107663A50MG
Encompass_Preferred
MIF-1 TFA (Pro-Leu-Gly-NH2 TFA) is an endogenous brain peptide supplied as the trifluoroacetic acid salt. It acts as a dopamine receptor allosteric modulator, crosses the blood-brain barrier, and is used in neuroscience and pharmacology research related to dopaminergic signaling, melanin formation, and opioid-mediated analgesia.
  • High purity (99.3%).
  • Provided as the trifluoroacetic acid salt.
  • Crosses the blood-brain barrier.
  • Molecular weight 398.38 and molecular formula C15H25F3N4O5.
  • Sealed storage: powder stable at -80°C (2 years) or -20°C (1 year).
  • For research use only.
Catalog No. 50-002-40306