Learn More
Enzo Life Sciences Mifepristone (100 mg). CAS: 84371-65-3
SDP
Supplier:  Enzo Life Sciences BMLS5100100
Encompass_Preferred
Progesterone receptor antagonist. Alternative name: RU 486. Purity: ≥99% (Assay). Solubility: Soluble in methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1747