Learn More
Selleck Chemical LLC Mifepristone S2606-1g
SDP
Supplier:  Selleck Chemical LLC S26061G
Encompass_Preferred
Mifepristone is a remarkably active antagonist of progesterone receptor and glucocorticoid receptor with IC50 of 0 2 nM and 2 6 nM respectively Mifepristone promotes cell autophagy and apoptosis decreases Bcl-2 level and increases Beclin1 level accompanied by weakened interaction between Bcl-2 and Beclin1 Mifepristone (RU486) can be used to induce animal models of Spontaneous Abortion
Catalog No. 50-311-2587