Learn More
Abcam Mifepristone (RU486), Progesterone and Glucocorticoid receptor antagonist, 500MG
Supplier:  Abcam AB120356500MG
Encompass_Preferred
MW 429.6 Da, Purity >99%. A cell-permeable synthetic steroid that acts as a potent selective antagonist of progesterone (PR) and glucocorticoid (GR) receptors in vitro and in vivo. Acts as a silent antagonist at PR and has a higher affinity than progesterone.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4619