Learn More
Medchemexpress LLC Mifepristone (Standard) | 84371-65-3 | 99.4% | 429.59 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13683R50MG
Encompass_Preferred
Mifepristone (Standard) is an analytical standard intended for research and analytical applications. It acts as a progesterone receptor (PR) and glucocorticoid receptor (GR) antagonist. This standard is commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Progesterone receptor (PR) and glucocorticoid receptor (GR) antagonist
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-003-48559