Learn More
Medchemexpress LLC Mifepristone (Standard) | 84371-65-3 | 99.42% | 429.59 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY13683R
Encompass_Preferred
Mifepristone (Standard) is an analytical standard of Mifepristone, designed for various research and analytical applications. It functions as a potent progesterone receptor (PR) and glucocorticoid receptor (GR) antagonist, exhibiting IC50s of 0.2 nM and 2.6 nM respectively in in vitro assays.
  • Intended for research and analytical applications
  • Functions as a progesterone receptor (PR) and glucocorticoid receptor (GR) antagonist
  • Effective in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS
Catalog No. 50-003-48830