Learn More
Cayman Chemical Mifepristoned3 GC MS Standards
SDP
Supplier:  Cayman Chemical 10010660500 UG
Encompass_Preferred
An internal standard for the quantification of mifepristone by GC- or LC-MS
Catalog No. 50-289-1276