Learn More
Medchemexpress LLC Migalastat hydrochloride (GR181413A) | 75172-81-5 | 99.7% | 199.63 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHY14929A
Encompass_Preferred
Migalastat hydrochloride (GR181413A) is an orally active α-galactosidase A molecular chaperone. It functions by binding to the active site of certain unstable mutant forms of α-galactosidase A, which facilitates their transport to the lysosome. Once in the acidic environment of the lysosome, Migalastat dissociates, allowing the mutant α-galactosidase A to exhibit biological activity.
  • Orally active α-galactosidase A molecular chaperone
  • IC50 value of 0.04 μM for human α-Gal A
  • Facilitates transport of unstable mutant α-galactosidase A to the lysosome
  • Enables mutant α-galactosidase A to exhibit biological activity
  • Reduces Gb3 accumulation and lysosome volume in EHK cells mutated α-Gal A
  • Increases α-Gal A activity in heart, kidney, spleen, and liver in transgenic mice
  • Reduces lyso-Gb3 levels in kidney, heart, and skin in transgenic mice
Catalog No. 50-003-64798