Learn More
Hoefer, Inc Hoefer™ SE250 Mighty Small II Mini Vertical Protein Electrophoresis Unit (Complete unit with 1.0mm)
SDP
Supplier:  Hoefer, Inc SE25010A1.0
Encompass_Preferred
Designed for quick screening and rapid separationFeatures:Maximum glass plate size 10 x 8 cmEfficient active cooling ensures sharp bandsQuick and easy to assembleRequires minimal bufferEasily upgradable to the SE260 (with optional accessories) resulting in the additional capability of accommodating both 10 x 8 and 10 x 10.5 cm gels, enabling a wider variety of precast gel optionsComplete Units Include:Buffer Chamber with Cooling Core; Safety Lid with High Voltage Leads; Glass Plates, Rectangular, 10 x 8 cm, 10 pcs; Alumina Plates, Notched, 10 x 8 cm, 2 pcs; Combs,10 well, 2 pcs; T-Spacers, 4 pcs; Gel casterBasic Unit Includes:The same as complete unit without the combs, spacers, and caster.Specifications:Glass Plate Size (W x H) 10 x 8 cmSpacers 0.75 mm, 1.0 mm, 1.5 mmMaximum Power Settings 500 V, 500 mA, 12 WMaximum Temperature 45°CIndoor Use 4 to 40°CHumidity Up to 80%Unit Dimension (W x H x D) 16.5 x 16 x 16 cm
Catalog No. NC2740682