Learn More
AdipoGen Miglitol (10 mg)
SDP
Supplier:  AdipoGen AGCR13635M010
Encompass_Preferred
Chemical. CAS: 72432-03-2. Formula: C8H17NO5. MW: 207.2. Anti-diabetic agent with antihyperglycemic activity. alpha-Glucosidase inhibitor. Inhibits lysosomal alpha-glucosidase, sucrase, maltase and isomaltase (IC50= 0.35, 0.11, 1.3, and 1.2µM, respectively) as well as maltase-glucoamylase (IC50 = 1.0µM). alpha-Glucosidase enzymes hydrolyze oligosaccharides and disaccharides into glucose and other monosaccharides. Inhibition by miglitol prevents the breakdown of larger carbohydrates into glucose. This suppresses postprandial hyperglycemia and reduces plasma glucose concentration in normal rats and in several animal models of diabetes without cardiovascular complications. Anti-obesity drug. Shown to inhibit adipogenesis of white adipocytes in vitro and to activate brown adipose tissue (BAT) in mice. Induces an enhanced and prolonged release of glucagon-like peptide-1, regulating appetite and stabilizing body weight in humans.
Catalog No. 50-205-4007