Learn More
Medchemexpress LLC HY-101266 5mg Medchemexpress, Milademetan CAS:1398568-47-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1012665MG
Encompass_Preferred
Medchemexpress, HY-101266 5mg Milademetan CAS:1398568-47-2 Mliademetan is a specific MDM2 inhibitor, a pharmaceutical composition for use in treating acute myeloid leukemia (AML). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2078