Learn More
Medchemexpress LLC Milademetan tosylate hydrate | 2095625-97-9 | 808.74 | C37H44Cl2FN5O8S | 1 ML

Supplier: Medchemexpress LLC HY101266B10MM 1ML
Milademetan tosylate hydrate is the tosylate salt hydrate of milademetan, used as a research reagent to study MDM2-p53 interactions, apoptosis, and related enzymatic targets. It is supplied for preparation of concentrated stock solutions for in vitro assays.
- CAS number: 2095625-97-9.
- Molecular weight: 808.74 g/mol.
- Chemical formula: C37H44Cl2FN5O8S.
- Form: tosylate hydrate (salt form).
- Targets: MDM2-p53 interaction and apoptosis-related enzymes.
- Solvent guidance: soluble in DMSO; example stock preparation 50 mg/mL.
- Stock concentrations: preparation guidance for 1 mM, 5 mM, 10 mM stocks.
- Storage: store sealed, protected from moisture and light; in solvent -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.