Learn More
Medchemexpress LLC Milademetan tosylate hydrate | 2095625-97-9 | 808.74 | C37H44Cl2FN5O8S
SDP
Supplier:  Medchemexpress LLC HY101266B50MG
Encompass_Preferred
Milademetan tosylate hydrate is the tosylate hydrate salt form of milademetan, an orally active MDM2 inhibitor supplied for research use. It is intended for preclinical studies investigating MDM2-p53 pathway modulation and compound screening.
  • Tosylate hydrate salt form of milademetan.
  • Selective MDM2 inhibitor suitable for preclinical research.
  • Supplied in small research quantities appropriate for screening and assay development.
  • Recommended storage: sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not validated for clinical applications.
Catalog No. 50-002-12445